Learn More
N-Desmethylclozapine, Tocris
5-HT2C antagonist
Manufacturer:  R&D Systems 1007/50
Quantity 50mg
 View more versions of this product
Catalog No. 10-075-0
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
>99%
312.8
N-Desmethylclozapine
C17H17ClN4
8-Chloro-11-(1-piperazinyl)-5H-dibenzo[b, e][1, 4]diazepine
Specifications
50mg
>99%
312.8
N-Desmethylclozapine
6104-71-8
C17H17ClN4
8-Chloro-11-(1-piperazinyl)-5H-dibenzo[b, e][1, 4]diazepine
Safety and Handling
Recommended Storage : Store at Room Temperature