Alfa Aesar™ N-(Diphenylmethylene)glycine ethyl ester, 98%

Manufacturer: Alfa Aesar™ B2558014

Quantity 25g

 View more versions of this product

Catalog No. AAB2558014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 69555-14-2
    Molecular Formula C17H17NO2
    Formula Weight 267.33
    MDL Number MFCD00010590
    Synonym Ethyl N-(diphenylmethylene)glycinate
    Chemical Name or Material N-(Diphenylmethylene)glycine ethyl ester

    Specifications

    Formula Weight 267.33
    Molecular Formula C17H17NO2
    MDL Number MFCD00010590
    CAS 69555-14-2
    Synonym Ethyl N-(diphenylmethylene)glycinate
    Melting Point 51° to 55°C
    Quantity 25g
    Flash Point >110°C (230°F)
    Chemical Name or Material N-(Diphenylmethylene)glycine ethyl ester
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit