Alfa Aesar™ N(epsilon)-Boc-L-lysine, 97%

Manufacturer: Alfa Aesar™ B2173806

Quantity 5g

 View more versions of this product

Catalog No. AAB2173806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2418-95-3
    Molecular Formula C11H22N2O4
    Formula Weight 246.31
    MDL Number MFCD00037221
    Beilstein 4252546
    Synonym N(epsilon)-tert-Butoxycarbonyl-L-lysine; H-Lys(Boc)-OH
    Chemical Name or Material N(epsilon)-Boc-L-lysine

    Specifications

    Formula Weight 246.31
    Molecular Formula C11H22N2O4
    MDL Number MFCD00037221
    CAS 2418-95-3
    Synonym N(epsilon)-tert-Butoxycarbonyl-L-lysine; H-Lys(Boc)-OH
    Melting Point ≈250°C (dec.)
    Quantity 5g
    Beilstein 4252546
    Chemical Name or Material N(epsilon)-Boc-L-lysine
    Assay 97%

    Description

    Applications
    It is used as therapy to treat recurrent herpes simplex infections.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit