Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethanoyl-D-erythro-sphingosine, synthetical, 99+%, ACROS Organics™

Click to view available options
Quantity:
50mg
Packaging:
Glass bottle
3102-57-6
C20H39NO3
341.536
MFCD00153903
BLTCBVOJNNKFKC-QUDYQQOWSA-N
n-acetylsphingosine,c2-ceramide,c2 ceramide,n-acetyl-sphing-4-enine,n-acetyl-ceramide,d-erythro-sphingosine, n-acetyl,n-acetyl-d-sphingosine,acetyl ceramide,c2 ceramide d18:1/2:0,acetamide, n-2-hydroxy-1-hydroxymethyl-3-heptadecenyl-, r-r*,s*-e
5497136
CHEBI:46979
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)C)O
3102-57-6
C20H39NO3
341.536
MFCD00153903
BLTCBVOJNNKFKC-QUDYQQOWSA-N
n-acetylsphingosine,c2-ceramide,c2 ceramide,n-acetyl-sphing-4-enine,n-acetyl-ceramide,d-erythro-sphingosine, n-acetyl,n-acetyl-d-sphingosine,acetyl ceramide,c2 ceramide d18:1/2:0,acetamide, n-2-hydroxy-1-hydroxymethyl-3-heptadecenyl-, r-r*,s*-e
5497136
CHEBI:46979
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)C)O
NMR Complies
Color White
Packaging Glass bottle
Quantity 50mg
Melting Point 93°C to 96°C (Clear point)
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Formula Weight 341.53
Physical Form Powder
Percent Purity ≥99.0%
Chemical Name or Material N-Ethanoyl-D-erythro-sphingosine, synthetical, 99+%
Show More Show Less