Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Ethyl-1-butylamine, 99%

Manufacturer:  Alfa Aesar™ B2288130

Quantity 250g

 View more versions of this product

Catalog No. AAB2288130


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13360-63-9
CH3(CH2)3NHCH2CH3
MFCD00009428
1731324
N-Ethyl-1-butylamine
C6H15N
101.19
UN2733
N-(n-Butyl)ethylamine

Specifications

13360-63-9
CH3(CH2)3NHCH2CH3
MFCD00009428
110°C
250g
Amine-like
N-Ethyl-1-butylamine
C6H15N
101.19
UN2733
1731324
12°C (54°F)
N-(n-Butyl)ethylamine
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318-H302
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

RTECSNumber : EO4880000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit