Alfa Aesar™ N-Ethyl-2-nitroaniline, 98%

Manufacturer: Alfa Aesar™ H5514803

Quantity 1g

 View more versions of this product

Catalog No. AAH5514803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10112-15-9
    Formula Weight 166.18
    MDL Number MFCD00007091
    Chemical Name or Material N-Ethyl-2-nitroaniline

    Specifications

    Formula Weight 166.18
    Refractive Index 1.6450
    MDL Number MFCD00007091
    Color Colorless to dark orange
    Physical Form Liquid
    Density 1.19g/mL
    CAS 10112-15-9
    Boiling Point 168°C (20mmHg)
    Quantity 1g
    Chemical Name or Material N-Ethyl-2-nitroaniline
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit