Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethylaniline 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
103-69-5
C8H11N
121.18
MFCD00009025
OJGMBLNIHDZDGS-UHFFFAOYSA-N
ethylaniline,n-ethyl aniline,benzenamine, n-ethyl,anilinoethane,n-ethylbenzenamine,ethylphenylamine,n-ethyl-n-phenylamine,aniline, n-ethyl,n-ethylaminobenzene,aethylanilin
7670
CHEBI:34870
N-ethylaniline
CCNC1=CC=CC=C1
103-69-5
C8H11N
121.18
MFCD00009025
OJGMBLNIHDZDGS-UHFFFAOYSA-N
ethylaniline,n-ethyl aniline,benzenamine, n-ethyl,anilinoethane,n-ethylbenzenamine,ethylphenylamine,n-ethyl-n-phenylamine,aniline, n-ethyl,n-ethylaminobenzene,aethylanilin
7670
CHEBI:34870
N-ethylaniline
CCNC1=CC=CC=C1
Melting Point -63°C
Flash Point 85°C
Boiling Point 205°C
Specific Gravity 0.963
Density 0.9630g/mL
Refractive Index 1.5530 to 1.555
Infrared Spectrum Authentic
Quantity 2.5kg
Packaging Glass bottle
Color Brown to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5NHC2H5
Beilstein 12, 159
Merck Index 15, 3819
Solubility Information Solubility in water: 50g/L water (20°C). Other solubilities: soluble in alcohol,ether and many other organic,solvents
Formula Weight 121.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material N-Ethylaniline, 98%
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-135-5

RTECSNumber : BX9780000

TSCA : TSCA