Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethylbenzylamine, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
14321-27-8
C9H13N
135.21
MFCD00009031
HVAAHUDGWQAAOJ-UHFFFAOYSA-N
n-ethylbenzylamine,benzenemethanamine, n-ethyl,ethylbenzylamine,n-benzylethylamine,benzyl ethyl amine,benzylamine, n-ethyl,benzylethylamine,unii-mm8324ke8v,n-benzyl-n-ethylamine,benzyl-ethyl-amine
84352
N-benzylethanamine
CCNCC1=CC=CC=C1
14321-27-8
C9H13N
135.21
MFCD00009031
HVAAHUDGWQAAOJ-UHFFFAOYSA-N
n-ethylbenzylamine,benzenemethanamine, n-ethyl,ethylbenzylamine,n-benzylethylamine,benzyl ethyl amine,benzylamine, n-ethyl,benzylethylamine,unii-mm8324ke8v,n-benzyl-n-ethylamine,benzyl-ethyl-amine
84352
N-benzylethanamine
CCNCC1=CC=CC=C1
Color Colorless to Yellow
Density 0.9000g/mL
Specific Gravity 0.9
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5100 to 1.5130
Flash Point 66°C
Quantity 100g
Boiling Point 191.0°C to 194.0°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula C6H5CH2NHC2H5
Beilstein 12,102
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ethyl ether and benzene
Formula Weight 135.21
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material N-Ethylbenzylamine, 95%
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

EINECSNumber : 238-265-1

TSCA : TSCA