Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethylcyclohexylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
5459-93-8
C8H17N
127.23
AGVKXDPPPSLISR-UHFFFAOYSA-N
n-ethylcyclohexylamine,cyclohexanamine, n-ethyl,n-cyclohexylethylamine,vulkacit hx,accelerator hx,cyclohexylamine, n-ethyl,ethylamino cyclohexane,cyclohexyl ethyl amine,n-ethyl cyclohexyl amine,unii-yjk13p0h3e
21609
N-ethylcyclohexanamine
CCNC1CCCCC1
5459-93-8
C8H17N
127.23
AGVKXDPPPSLISR-UHFFFAOYSA-N
n-ethylcyclohexylamine,cyclohexanamine, n-ethyl,n-cyclohexylethylamine,vulkacit hx,accelerator hx,cyclohexylamine, n-ethyl,ethylamino cyclohexane,cyclohexyl ethyl amine,n-ethyl cyclohexyl amine,unii-yjk13p0h3e
21609
N-ethylcyclohexanamine
CCNC1CCCCC1
Specific Gravity 0.85
Flash Point 30°C
Boiling Point 164°C
Color Colorless to Yellow
Density 0.8500g/mL
Packaging Glass bottle
Water 0.5% max.
Refractive Index 1.4500 to 1.4530 (20°C, 589nm)
Quantity 100mL
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 127.23
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material N-Ethylcyclohexylamine
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 226-733-8