Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-ethylglycine 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
627-01-0
C4H9NO2
103.121
MFCD00037794
YPIGGYHFMKJNKV-UHFFFAOYSA-N
n-ethylglycine,2-ethylamino acetic acid,n-ethyl glycine,etgly,glycine, n-ethyl,monoethylglycine,pubchem23122,acmc-1au6b,ypiggyhfmkjnkv-uhfffaoysa
316542
CHEBI:15620
2-(ethylamino)acetic acid
CCNCC(=O)O
627-01-0
C4H9NO2
103.121
MFCD00037794
YPIGGYHFMKJNKV-UHFFFAOYSA-N
n-ethylglycine,2-ethylamino acetic acid,n-ethyl glycine,etgly,glycine, n-ethyl,monoethylglycine,pubchem23122,acmc-1au6b,ypiggyhfmkjnkv-uhfffaoysa
316542
CHEBI:15620
2-(ethylamino)acetic acid
CCNCC(=O)O
Infrared Spectrum Authentic
Color White
Melting Point 182°C to 186°C
Quantity 1g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CH2NHCH2COOH
Formula Weight 103.12
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material N-Ethylglycine
Show More Show Less