Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Ethylheptylamine, 97%

Click to view available options
Quantity:
1g
5g
25g
66793-76-8
C9H21N
143.274
MFCD00049012
IUZZLNVABCISOI-UHFFFAOYSA-N
n-ethylheptylamine,n-ethyl-1-heptanamine,1-heptanamine, n-ethyl,ethylheptylamine,ethyl heptyl amine,ethylheptyl-amine,n-ethylheptanamine,n-ethyl-n-heptylamine,n-heptyl-n-ethylamine,acmc-209nwl
48038
N-ethylheptan-1-amine
CCCCCCCNCC
66793-76-8
C9H21N
143.274
MFCD00049012
IUZZLNVABCISOI-UHFFFAOYSA-N
n-ethylheptylamine,n-ethyl-1-heptanamine,1-heptanamine, n-ethyl,ethylheptylamine,ethyl heptyl amine,ethylheptyl-amine,n-ethylheptanamine,n-ethyl-n-heptylamine,n-heptyl-n-ethylamine,acmc-209nwl
48038
N-ethylheptan-1-amine
CCCCCCCNCC
Color Yellow
Quantity 25g
Boiling Point 181°C
Density 0.772g/mL
Refractive Index 1.4240
Linear Formula CH3CH2NH(CH2)6CH3
UN Number UN2733
Beilstein 1733571
Formula Weight 143.27
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material N-Ethylheptylamine
Show More Show Less

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.