Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Ethylthiourea, 99%

Manufacturer:  Alfa Aesar™ B2476109

Quantity 10g

 View more versions of this product

Catalog No. AAB2476109


  Check Availability
Add to cart

Specifications

Description

Applications
N-Ethylthiourea was used in the preparation of Re (III) complexes, which are precursors for the new rhenium complexes, potentially useful in nuclear medicine. It was also used in two-step protocol for the first chemoselective cleavage of 2-hydroxy acid amides

Solubility
Soluble in water (24g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

625-53-6
104.17
UN2811
N-Ethylthiourea
C3H8N2S
MFCD00004939
1699551

Specifications

625-53-6
104.17
MFCD00004939
1699551
N-Ethylthiourea
C3H8N2S
108° to 112°C
UN2811
10g
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : YT3700000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit