Alfa Aesar™ N-Ethylthiourea, 99%

Manufacturer: Alfa Aesar™ B2476130

Quantity 250g

 View more versions of this product

Catalog No. AAB2476130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 625-53-6
    Molecular Formula C3H8N2S
    Formula Weight 104.17
    MDL Number MFCD00004939
    UN Number UN2811
    Beilstein 1699551
    Chemical Name or Material N-Ethylthiourea

    Specifications

    Formula Weight 104.17
    TSCA TSCA
    Molecular Formula C3H8N2S
    Melting Point 108° to 112°C
    MDL Number MFCD00004939
    CAS 625-53-6
    UN Number UN2811
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P280f-P301+P310-P330-P501a
    Wear protective clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number YT3700000
    Beilstein 1699551
    Health Hazard 1 Danger
    Chemical Name or Material N-Ethylthiourea
    Assay 99%

    Description

    Applications
    N-Ethylthiourea was used in the preparation of Re (III) complexes, which are precursors for the new rhenium complexes, potentially useful in nuclear medicine. It was also used in two-step protocol for the first chemoselective cleavage of 2-hydroxy acid amides

    Solubility
    Soluble in water (24g/L).

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P280f-P301+P310-P330-P501a
    Wear protective clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit