Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Ethylthiourea, 99%

Click to view available options
Quantity:
10g
50g
250g
625-53-6
C3H8N2S
104.171
MFCD00004939
GMEHFXXZSWDEDB-UHFFFAOYSA-N
n-ethylthiourea,1-ethylthiourea,1-ethyl-2-thiourea,thiourea, ethyl,n-ethylthiocarbamide,ethyl thiourea,ethyl-2-thiourea,thiourea, n-ethyl,urea, 1-ethyl-2-thio,unii-u54or2e97e
3032338
ethylthiourea
CCNC(=S)N
625-53-6
C3H8N2S
104.171
MFCD00004939
GMEHFXXZSWDEDB-UHFFFAOYSA-N
n-ethylthiourea,1-ethylthiourea,1-ethyl-2-thiourea,thiourea, ethyl,n-ethylthiocarbamide,ethyl thiourea,ethyl-2-thiourea,thiourea, n-ethyl,urea, 1-ethyl-2-thio,unii-u54or2e97e
3032338
ethylthiourea
CCNC(=S)N
Applications
N-Ethylthiourea was used in the preparation of Re (III) complexes, which are precursors for the new rhenium complexes, potentially useful in nuclear medicine. It was also used in two-step protocol for the first chemoselective cleavage of 2-hydroxy acid amides

Solubility
Soluble in water (24g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Quantity 50g
Melting Point 108°C to 112°C
UN Number UN2811
Beilstein 1699551
Formula Weight 104.17
Percent Purity 99%
Chemical Name or Material N-Ethylthiourea

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : YT3700000

TSCA : TSCA