Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-2-cyclohexyl-D-glycine, 95%

Click to view available options
Quantity:
250mg
1g
5g
198543-96-3
C23H25NO4
379.456
MFCD00190877
BWQQGHPODCJZDB-OAQYLSRUSA-N
fmoc-d-chg-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid,r-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid,ambotzfaa1666,bicr146,r-a-fmoc-amino-cyclohexaneacetic acid,r-alpha-fmoc-amino cyclohexaneacetic acid,n-alpha-9-fluorenylmethyloxycarbonyl-d-cyclohexylglycine,2r-2-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino acetic acid
6976355
(2R)-2-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
198543-96-3
C23H25NO4
379.456
MFCD00190877
BWQQGHPODCJZDB-OAQYLSRUSA-N
fmoc-d-chg-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid,r-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid,ambotzfaa1666,bicr146,r-a-fmoc-amino-cyclohexaneacetic acid,r-alpha-fmoc-amino cyclohexaneacetic acid,n-alpha-9-fluorenylmethyloxycarbonyl-d-cyclohexylglycine,2r-2-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino acetic acid
6976355
(2R)-2-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 379.46
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material N-Fmoc-2-cyclohexyl-D-glycine

storageNote1 : -30° to -10°C