Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-2-fluoro-L-phenylalanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
205526-26-7
C24H20FNO4
405.425
MFCD00672552
ARHOAMSIDCQWEW-QFIPXVFZSA-N
fmoc-phe 2-f-oh,fmoc-l-2-fluorophenylalanine,fmoc-2-fluoro-l-phenylalanine,fmoc-l-2-fluorophe,fmoc-2-fluoro-l-phe,fmoc-l-phe 2-f-oh,fmoc-l-2-fluoro-phe-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl propanoic acid,s-n-fmoc-2-fluorophenylalanine,n-alpha-9-fluorenylmethyloxycarbonyl-l-2-fluoro-phenylalanine
2734535
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)propanoic acid
C1=CC=C(C(=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)F
205526-26-7
C24H20FNO4
405.425
MFCD00672552
ARHOAMSIDCQWEW-QFIPXVFZSA-N
fmoc-phe 2-f-oh,fmoc-l-2-fluorophenylalanine,fmoc-2-fluoro-l-phenylalanine,fmoc-l-2-fluorophe,fmoc-2-fluoro-l-phe,fmoc-l-phe 2-f-oh,fmoc-l-2-fluoro-phe-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl propanoic acid,s-n-fmoc-2-fluorophenylalanine,n-alpha-9-fluorenylmethyloxycarbonyl-l-2-fluoro-phenylalanine
2734535
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)propanoic acid
C1=CC=C(C(=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)F
Beilstein 9437460
Formula Weight 405.43
Physical Form Powder
Percent Purity 95%
Color White
Quantity 250mg
Chemical Name or Material N-Fmoc-2-fluoro-L-phenylalanine

storageNote1 : 2° to 8°C