Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-2-octyl-L-glycine, 98%

Click to view available options
Quantity:
250mg
1g
5g
193885-59-5
C25H31NO4
409.526
MFCD01311753
LSMLSLHVRMSYPT-QHCPKHFHSA-N
fmoc-octyl-gly-oh,fmoc-2s-2-amino-decanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid,ambotzfaa1598,fmoc-s-octylglycine,s-n-fmoc-octylglycine-oh,n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine,2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid,s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid,2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
23257324
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid
CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
193885-59-5
C25H31NO4
409.526
MFCD01311753
LSMLSLHVRMSYPT-QHCPKHFHSA-N
fmoc-octyl-gly-oh,fmoc-2s-2-amino-decanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid,ambotzfaa1598,fmoc-s-octylglycine,s-n-fmoc-octylglycine-oh,n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine,2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid,s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid,2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
23257324
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid
CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Applications
A homolog of lipophilic building block for SAR studies.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Formula Weight 409.53
Physical Form Powder
Percent Purity 98%
Color White
Melting Point 140°C to 143°C
Quantity 1g
Chemical Name or Material N-Fmoc-2-octyl-L-glycine