Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-3-(2-naphthyl)-D-alanine, 95%

Manufacturer:  Alfa Aesar™ H51974MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51974MD


  Check Availability
Add to cart

Specifications

Description

Applications
N-Fmoc-3-(2-naphthyl)-D-alanine is an Fmoc-protected alanine derivative and an intermediate used in the preparation of phosphoserine-containing tetrapeptides with hydrophobic N-terminal acyl groups as inhibitors of the BRCA1 protein.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

138774-94-4
437.49
7864489
N-Fmoc-3-(2-naphthyl)-D-alanine
C28H23NO4
MFCD00077044
Fmoc-D-2-Nal-OH

Specifications

138774-94-4
437.49
7864489
Fmoc-D-2-Nal-OH
95%
C28H23NO4
MFCD00077044
250mg
N-Fmoc-3-(2-naphthyl)-D-alanine
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit