Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-3-(2-pyridyl)-L-alanine, 95%

Click to view available options
Quantity:
250mg
1g
185379-40-2
C23H20N2O4
388.423
MFCD00672564
DXIVJCDZOMUITR-NRFANRHFSA-N
fmoc-l-2-pyridylalanine,fmoc-2-pal-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-2-yl propanoic acid,fmoc-2'-pyridyl-l-ala,fmoc-beta-2-pyridyl-ala-oh,s-n-fmoc-2-pyridyl alanine,fmoc-l-3-2-pyridyl-alanine,fmoc-l-3-2-pyridyl alanine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-2-yl propanoic acid,fmoc-d-3-2-pyridyl alanine
978320
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-2-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CC=N4)C(=O)O
185379-40-2
C23H20N2O4
388.423
MFCD00672564
DXIVJCDZOMUITR-NRFANRHFSA-N
fmoc-l-2-pyridylalanine,fmoc-2-pal-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-2-yl propanoic acid,fmoc-2'-pyridyl-l-ala,fmoc-beta-2-pyridyl-ala-oh,s-n-fmoc-2-pyridyl alanine,fmoc-l-3-2-pyridyl-alanine,fmoc-l-3-2-pyridyl alanine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-2-yl propanoic acid,fmoc-d-3-2-pyridyl alanine
978320
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-2-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CC=N4)C(=O)O
Formula Weight 388.42
Physical Form Powder
Percent Purity 95%
Color Brown
Quantity 1g
Chemical Name or Material N-Fmoc-3-(2-pyridyl)-L-alanine

storageNote1 : 2° to 8°C