Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-3,3-diphenyl-L-alanine, 95%

Manufacturer:  Alfa Aesar™ H52176MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52176MD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

201484-50-6
463.53
UN3077
Fmoc-beta-phenyl-Phe-OH; Fmoc-beta,beta-diphenyl-Ala-OH
C30H25NO4
MFCD00672337
8873562
N-Fmoc-3,3-diphenyl-L-alanine

Specifications

201484-50-6
463.53
MFCD00672337
8873562
Fmoc-beta-phenyl-Phe-OH; Fmoc-beta,beta-diphenyl-Ala-OH
95%
C30H25NO4
125° to 129°C
UN3077
250mg
N-Fmoc-3,3-diphenyl-L-alanine

Safety and Handling

SDS & Safety Documents

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit