Alfa Aesar™ N-Fmoc-3-(3-thienyl)-L-alanine, 95%

Manufacturer: Alfa Aesar™ H5216706

Quantity 5g

 View more versions of this product

Catalog No. AAH5216706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 186320-06-9
    Molecular Formula C22H19NO4S
    Formula Weight 393.46
    MDL Number MFCD00151916
    Beilstein 9512976
    Synonym Fmoc-beta-(3-thienyl)-Ala-OH
    Chemical Name or Material N-Fmoc-3-(3-thienyl)-L-alanine

    Specifications

    Formula Weight 393.46
    Molecular Formula C22H19NO4S
    MDL Number MFCD00151916
    CAS 186320-06-9
    Beilstein 9512976
    Quantity 5g
    Synonym Fmoc-beta-(3-thienyl)-Ala-OH
    Chemical Name or Material N-Fmoc-3-(3-thienyl)-L-alanine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit