Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-3-(4-pyridyl)-D-alanine, 95%

Manufacturer:  Alfa Aesar™ H5219306

Quantity 5g

 View more versions of this product

Catalog No. AAH5219306


  Check Availability
Add to cart

Specifications

Description

Applications
N-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

205528-30-9
388.42
Fmoc-beta-(4-pyridyl)-D-Ala-OH
C23H20N2O4
MFCD00672567
N-Fmoc-3-(4-pyridyl)-D-alanine

Specifications

205528-30-9
388.42
5g
N-Fmoc-3-(4-pyridyl)-D-alanine
C23H20N2O4
MFCD00672567
Fmoc-beta-(4-pyridyl)-D-Ala-OH
95%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit