Alfa Aesar™ N-Fmoc-3-(4-pyridyl)-D-alanine, 95%

Manufacturer: Alfa Aesar™ H52193MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52193MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 205528-30-9
    Molecular Formula C23H20N2O4
    Formula Weight 388.42
    MDL Number MFCD00672567
    Synonym Fmoc-beta-(4-pyridyl)-D-Ala-OH
    Chemical Name or Material N-Fmoc-3-(4-pyridyl)-D-alanine

    Specifications

    Formula Weight 388.42
    MDL Number MFCD00672567
    Quantity 250mg
    Molecular Formula C23H20N2O4
    CAS 205528-30-9
    Synonym Fmoc-beta-(4-pyridyl)-D-Ala-OH
    Chemical Name or Material N-Fmoc-3-(4-pyridyl)-D-alanine
    Assay 95%

    Description

    Applications
    N-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit