Alfa Aesar™ N-Fmoc-3-(4-thiazolyl)-L-alanine, 95%

Manufacturer: Alfa Aesar™ H5207106

Quantity 5g

 View more versions of this product

Catalog No. AAH5207106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 205528-32-1
    Molecular Formula C21H18N2O4S
    Formula Weight 394.44
    MDL Number MFCD00672568
    Synonym Fmoc-beta-(4-thiazolyl)-Ala-OH
    Chemical Name or Material N-Fmoc-3-(4-thiazolyl)-L-alanine

    Specifications

    Formula Weight 394.44
    Molecular Formula C21H18N2O4S
    Melting Point 178° to 179°C
    MDL Number MFCD00672568
    CAS 205528-32-1
    Quantity 5g
    Synonym Fmoc-beta-(4-thiazolyl)-Ala-OH
    Chemical Name or Material N-Fmoc-3-(4-thiazolyl)-L-alanine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit