Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-4-benzoyl-D-phenylalanine, 98%

Click to view available options
Quantity:
250mg
1g
5g
117666-97-4
C31H25NO5
491.543
MFCD00237666
SYOBJKCXNRQOGA-MUUNZHRXSA-N
fmoc-d-bpa-oh,fmoc-4-benzoyl-d-phenylalanine,fmoc-d-4-benzoylphenylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzoylphenyl propanoic acid,2r-3-4-benzoylphenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,ambotzfaa1665,boc-p-bz-d-phe-oh,fmoc-p-bz-d-phe-oh,fmoc-d-bpa-oh hplc,n-alpha-9-fluorenylmethyloxycarbonyl-4-benzoyl-d-phenylalanine
14233361
(2R)-3-(4-benzoylphenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
117666-97-4
C31H25NO5
491.543
MFCD00237666
SYOBJKCXNRQOGA-MUUNZHRXSA-N
fmoc-d-bpa-oh,fmoc-4-benzoyl-d-phenylalanine,fmoc-d-4-benzoylphenylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzoylphenyl propanoic acid,2r-3-4-benzoylphenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,ambotzfaa1665,boc-p-bz-d-phe-oh,fmoc-p-bz-d-phe-oh,fmoc-d-bpa-oh hplc,n-alpha-9-fluorenylmethyloxycarbonyl-4-benzoyl-d-phenylalanine
14233361
(2R)-3-(4-benzoylphenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
Formula Weight 491.54
Physical Form Powder
Percent Purity 98%
Color Brown
Quantity 250mg
Chemical Name or Material N-Fmoc-4-benzoyl-D-phenylalanine

storageNote1 : 2° to 8°C