Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-4-fluoro-D-phenylalanine, 95%

Click to view available options
Quantity:
1g
5g
177966-64-2
C24H20FNO4
405.425
MFCD00235900
IXUMACXMEZBPJG-JOCHJYFZSA-N
fmoc-d-phe 4-f-oh,fmoc-d-4-fluorophenylalanine,fmoc-4-fluoro-d-phenylalanine,fmoc-d-4-fluorophe,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-fluorophenyl propanoic acid,fmoc-4-fluoro-d-phe,r-n-fmoc-4-fluorophenylalanine,fmoc-p-fluoro-d-phe-oh,n-alpha-9-fluorenylmethyloxycarbonyl-4-fluoro-d-phenylalanine,d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-fluoro
16213164
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-fluorophenyl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)F)C(=O)O
177966-64-2
C24H20FNO4
405.425
MFCD00235900
IXUMACXMEZBPJG-JOCHJYFZSA-N
fmoc-d-phe 4-f-oh,fmoc-d-4-fluorophenylalanine,fmoc-4-fluoro-d-phenylalanine,fmoc-d-4-fluorophe,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-fluorophenyl propanoic acid,fmoc-4-fluoro-d-phe,r-n-fmoc-4-fluorophenylalanine,fmoc-p-fluoro-d-phe-oh,n-alpha-9-fluorenylmethyloxycarbonyl-4-fluoro-d-phenylalanine,d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-fluoro
16213164
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-fluorophenyl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)F)C(=O)O
Formula Weight 405.43
Physical Form Powder
Percent Purity 95%
Color White
Quantity 1g
Chemical Name or Material N-Fmoc-4-fluoro-D-phenylalanine

storageNote1 : 2° to 8°C