Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-4-iodo-D-phenylalanine, 98%

Click to view available options
Quantity:
250mg
1g
5g
205526-29-0
C24H20INO4
513.331
MFCD00672557
LXOXXTQKKRJNNB-JOCHJYFZSA-N
fmoc-d-phe 4-i-oh,fmoc-4-iodo-d-phenylalanine,fmoc-d-4-iodophenylalanine,fmoc-d-4-iodophe,fmoc-4'-iodo-d-phe,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid,ambotzfaa1584,fmoc-4-iodo-d-phe-oh,fmoc-d-phe 4-i-oh hplc
16213204
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-iodophenyl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)I)C(=O)O
205526-29-0
C24H20INO4
513.331
MFCD00672557
LXOXXTQKKRJNNB-JOCHJYFZSA-N
fmoc-d-phe 4-i-oh,fmoc-4-iodo-d-phenylalanine,fmoc-d-4-iodophenylalanine,fmoc-d-4-iodophe,fmoc-4'-iodo-d-phe,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid,ambotzfaa1584,fmoc-4-iodo-d-phe-oh,fmoc-d-phe 4-i-oh hplc
16213204
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-iodophenyl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)I)C(=O)O
Applications
N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Sparingly Soluble in water (6.7E-4 g/L) (25°C).

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Beilstein 9159551
Solubility Information Sparingly Soluble in water (6.7E-4g/L) (25°C).
Formula Weight 513.32
Physical Form Solid
Percent Purity 98%
Quantity 250mg
Sensitivity Light sensitive
Chemical Name or Material N-Fmoc-4-iodo-D-phenylalanine

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold