Alfa Aesar™ N-Fmoc-4-methoxy-L-phenylalanine, 95%

Manufacturer: Alfa Aesar™ H52117MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52117MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 77128-72-4
    Molecular Formula C25H23NO5
    Formula Weight 417.45
    MDL Number MFCD00153368
    Synonym Fmoc-Tyr(Me)-OH; N-Fmoc-O-methyl-L-tyrosine
    Chemical Name or Material N-Fmoc-4-methoxy-L-phenylalanine

    Specifications

    Formula Weight 417.45
    Molecular Formula C25H23NO5
    MDL Number MFCD00153368
    Quantity 250mg
    Synonym Fmoc-Tyr(Me)-OH; N-Fmoc-O-methyl-L-tyrosine
    CAS 77128-72-4
    Chemical Name or Material N-Fmoc-4-methoxy-L-phenylalanine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit