Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-4-(trifluoromethyl)-L-phenylalanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
247113-86-6
C25H20F3NO4
455.433
MFCD00797581
YMEGJWTUWMVZPD-QFIPXVFZSA-N
fmoc-phe 4-cf3-oh,fmoc-l-4-trifluoromethylphenylalanine,fmoc-4-trifluoromethyl-l-phenylalanine,fmoc-l-4-trifluoromethylphe,4-trifluoromethyl-l-phenyl alanine, fmoc protected,4-trifluoromethyl-l-phenylalanine, n-fmoc protected,2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-trifluoromethyl phenyl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-trifluoromethyl phenyl propanoic acid,n-9h-fluoren-9-yl methoxy carbonyl-4-trifluoromethyl-l-phenylalanine
7021069
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-(trifluoromethyl)phenyl]propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)C(F)(F)F)C(=O)O
247113-86-6
C25H20F3NO4
455.433
MFCD00797581
YMEGJWTUWMVZPD-QFIPXVFZSA-N
fmoc-phe 4-cf3-oh,fmoc-l-4-trifluoromethylphenylalanine,fmoc-4-trifluoromethyl-l-phenylalanine,fmoc-l-4-trifluoromethylphe,4-trifluoromethyl-l-phenyl alanine, fmoc protected,4-trifluoromethyl-l-phenylalanine, n-fmoc protected,2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-trifluoromethyl phenyl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-trifluoromethyl phenyl propanoic acid,n-9h-fluoren-9-yl methoxy carbonyl-4-trifluoromethyl-l-phenylalanine
7021069
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-(trifluoromethyl)phenyl]propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)C(F)(F)F)C(=O)O
Formula Weight 455.43
Physical Form Powder
Percent Purity 95%
Color Brown
Melting Point 127°C to 130°C
Quantity 5g
Chemical Name or Material N-Fmoc-4-(trifluoromethyl)-L-phenylalanine

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold