Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-D-aspartic acid 4-allyl ester, 95%

Click to view available options
Quantity:
250mg
1g
5g
177609-12-0
C22H21NO6
395.411
MFCD01074688
FBNFRRNBFASDKS-LJQANCHMSA-N
fmoc-d-asp oall-oh,n-alpha-9-fluorenylmethyloxycarbonyl-d-aspartic acid beta allyl ester,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid,r-2-9h-fluoren-9-yl methoxy carbonylamino-4-allyloxy-4-oxobutanoic acid,d-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-, 4-2-propenyl ester,2r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid
10872947
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid
C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
177609-12-0
C22H21NO6
395.411
MFCD01074688
FBNFRRNBFASDKS-LJQANCHMSA-N
fmoc-d-asp oall-oh,n-alpha-9-fluorenylmethyloxycarbonyl-d-aspartic acid beta allyl ester,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid,r-2-9h-fluoren-9-yl methoxy carbonylamino-4-allyloxy-4-oxobutanoic acid,d-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-, 4-2-propenyl ester,2r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid
10872947
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid
C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Formula Weight 395.41
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material N-Fmoc-D-aspartic acid 4-allyl ester

storageNote1 : -30° to -10°C