Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-D-aspartic acid 4-benzyl ester, 95%

missing translation for 'orderingAttributeHoverText'
Quantity:
250mg
1g
5g
150009-58-8
C26H23NO6
445.471
MFCD00273445
OQGAELAJEGGNKG-HSZRJFAPSA-N
fmoc-d-asp obzl-oh,2r-4-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid,fmoc-l-aspartic acid-beta benzyl ester,d-aspartic acid,n-9h-fluoren-9-ylmethoxy carbonyl-, 4-phenylmethyl ester
40428625
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-phenylmethoxybutanoic acid
C1=CC=C(C=C1)COC(=O)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
150009-58-8
C26H23NO6
445.471
MFCD00273445
OQGAELAJEGGNKG-HSZRJFAPSA-N
fmoc-d-asp obzl-oh,2r-4-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid,fmoc-l-aspartic acid-beta benzyl ester,d-aspartic acid,n-9h-fluoren-9-ylmethoxy carbonyl-, 4-phenylmethyl ester
40428625
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-phenylmethoxybutanoic acid
C1=CC=C(C=C1)COC(=O)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 445.47
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material N-Fmoc-D-aspartic acid 4-benzyl ester

storageNote1 : -30° to -10°C