Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-D-glutamic acid 5-benzyl ester, 95%

Click to view available options
Quantity:
250mg
1g
5g
104091-11-4
C27H25NO6
459.498
MFCD00273449
HJJURMMMGPQIQP-XMMPIXPASA-N
fmoc-d-glu obzl-oh,fmoc-d-glu bzl-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid,2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid,ambotzfaa1485,fmoc-d-glutamic acid,a-benzyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamic-acid-gamma-benzyl ester,2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxycarbonylamino-5-oxo-pentanoic acid
10837618
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
104091-11-4
C27H25NO6
459.498
MFCD00273449
HJJURMMMGPQIQP-XMMPIXPASA-N
fmoc-d-glu obzl-oh,fmoc-d-glu bzl-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid,2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid,ambotzfaa1485,fmoc-d-glutamic acid,a-benzyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamic-acid-gamma-benzyl ester,2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxycarbonylamino-5-oxo-pentanoic acid
10837618
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 459.5
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material N-Fmoc-D-glutamic acid 5-benzyl ester

storageNote1 : -30° to -10°C