Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-D-glutamine, 95%

Click to view available options
Quantity:
5g
25g
112898-00-7
C20H20N2O5
368.389
MFCD00062959
IZKGGDFLLNVXNZ-QGZVFWFLSA-N
fmoc-d-glutamine,fmoc-d-gln-oh,fmoc-d-gln,n-a-fmoc-d-glutamine,n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamine,ambotzfaa1483,n-alpha-fmoc-d-glutamine,fmoc-d-gln-oh,n-alpha-9-fluorenylmethoxycarbonyl-d-glutamine,2r-5-amino-2-9h-fluoren-9-ylmethoxy oxo methyl amino-5-oxopentanoic acid
6992536
(2R)-5-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)N)C(=O)O
112898-00-7
C20H20N2O5
368.389
MFCD00062959
IZKGGDFLLNVXNZ-QGZVFWFLSA-N
fmoc-d-glutamine,fmoc-d-gln-oh,fmoc-d-gln,n-a-fmoc-d-glutamine,n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamine,ambotzfaa1483,n-alpha-fmoc-d-glutamine,fmoc-d-gln-oh,n-alpha-9-fluorenylmethoxycarbonyl-d-glutamine,2r-5-amino-2-9h-fluoren-9-ylmethoxy oxo methyl amino-5-oxopentanoic acid
6992536
(2R)-5-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)N)C(=O)O
Formula Weight 368.39
Physical Form Powder
Percent Purity 95%
Color White
Quantity 5g
Chemical Name or Material N-Fmoc-D-glutamine

storageNote1 : 2° to 8°C