Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-D-homophenylalanine, 98%

Click to view available options
Quantity:
1g
5g
135944-09-1
C25H23NO4
401.462
MFCD00151917
CIHPCIUGLIZADU-HSZRJFAPSA-N
fmoc-d-homophe-oh,fmoc-d-homophenylalanine,fmoc-d-hophe-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid,fmoc-d-hphe-oh,fmoc-d-hph-oh,fmoc-d-homophenyl-ala,fmoc-d-homophe,fmoc-d-hfe-oh,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid
7129818
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid
C1=CC=C(C=C1)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
135944-09-1
C25H23NO4
401.462
MFCD00151917
CIHPCIUGLIZADU-HSZRJFAPSA-N
fmoc-d-homophe-oh,fmoc-d-homophenylalanine,fmoc-d-hophe-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid,fmoc-d-hphe-oh,fmoc-d-hph-oh,fmoc-d-homophenyl-ala,fmoc-d-homophe,fmoc-d-hfe-oh,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid
7129818
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid
C1=CC=C(C=C1)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Beilstein 4847668
Formula Weight 401.46
Physical Form Powder
Percent Purity 98%
Color White
Quantity 5g
Chemical Name or Material N-Fmoc-D-homophenylalanine

storageNote1 : 2° to 8°C