Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-D-leucine, 98%

Manufacturer:  Alfa Aesar™ H2904103

Quantity 1g

 View more versions of this product

Catalog No. AAH2904103


  Check Availability
Add to cart

Specifications

Description

Applications
A convergent synthesis of the proposed structure of (+)-pestalazine B has been achieved in 4 steps using the N-alkylation of an unprotected tryptophan diketopiperazine with a 3a-bromopyrrolidinoindoline where the condensation between L-tryptophan methyl ester and N-Fmoc-D-leucine in the presence of EDC as coupling agent afforded the corresponding L-Trp-D-Leu dipeptide.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

114360-54-2
353.42
N-(9-Fluorenylmethoxy)carbonyl-D-leucine; Fmoc-D-Leu-OH
C21H23NO4
MFCD00062957
N-Fmoc-D-leucine

Specifications

114360-54-2
353.42
MFCD00062957
N-(9-Fluorenylmethoxy)carbonyl-D-leucine; Fmoc-D-Leu-OH
N-Fmoc-D-leucine
C21H23NO4
151° to 154°C
1g
+25° (c=1 in DMF)
98%

Safety and Handling

SDS & Safety Documents

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit