Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-D-norleucine, 95%

Click to view available options
Quantity:
5g
25g
112883-41-7
C21H23NO4
353.418
MFCD00155639
VCFCFPNRQDANPN-LJQANCHMSA-N
fmoc-d-nle-oh,fmoc-d-norleucine,r-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid,d-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,n-fmoc-d-norleucine,pubchem14959,d-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl,fmoc-d-nle-oh,n-alpha-9-fluorenylmethyloxycarbonyl-d-norleucine
2756111
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
112883-41-7
C21H23NO4
353.418
MFCD00155639
VCFCFPNRQDANPN-LJQANCHMSA-N
fmoc-d-nle-oh,fmoc-d-norleucine,r-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid,d-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,n-fmoc-d-norleucine,pubchem14959,d-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl,fmoc-d-nle-oh,n-alpha-9-fluorenylmethyloxycarbonyl-d-norleucine
2756111
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Beilstein 8227505
Formula Weight 353.42
Physical Form Powder
Percent Purity 95%
Color White
Quantity 25g
Chemical Name or Material N-Fmoc-D-norleucine

storageNote1 : 2° to 8°C