Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-D-phenylalanine, 98%

Click to view available options
Quantity:
1g
5g
25g
86123-10-6
C24H21NO4
387.435
MFCD00062955
SJVFAHZPLIXNDH-JOCHJYFZSA-N
fmoc-d-phenylalanine,fmoc-d-phe-oh,n-fmoc-d-phenylalanine,n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid,fmoc-r-beta-phenylalanine,n-9-fluorenylmethoxycarbonyl-d-phenylalanine,d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl,n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid
978332
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
86123-10-6
C24H21NO4
387.435
MFCD00062955
SJVFAHZPLIXNDH-JOCHJYFZSA-N
fmoc-d-phenylalanine,fmoc-d-phe-oh,n-fmoc-d-phenylalanine,n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid,fmoc-r-beta-phenylalanine,n-9-fluorenylmethoxycarbonyl-d-phenylalanine,d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl,n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid
978332
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Melting Point 181°C to 185°C
Optical Rotation +40° (c=1 in DMF)
Quantity 1g
Formula Weight 387.44
Percent Purity 98%
Chemical Name or Material N-Fmoc-D-phenylalanine

Recommended Storage : 2° to 8°C