Alfa Aesar™ N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water

Manufacturer: Alfa Aesar™ H2850006

Quantity 5g

 View more versions of this product

Catalog No. AAH2850006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 101555-62-8
    Molecular Formula C20H19NO4
    Formula Weight 337.38
    MDL Number MFCD00077067
    Synonym N-(9-Fluorenylmethoxy)carbonyl-D-proline; Fmoc-D-Pro-OH
    Chemical Name or Material N-Fmoc-D-proline

    Specifications

    Formula Weight 337.38
    Melting Point 110° to 116°C
    CAS 101555-62-8
    Synonym N-(9-Fluorenylmethoxy)carbonyl-D-proline; Fmoc-D-Pro-OH
    Molecular Formula C20H19NO4
    MDL Number MFCD00077067
    Quantity 5g
    Recommended Storage 2° to 8°C
    Chemical Name or Material N-Fmoc-D-proline
    Assay ≥98%, may contain up to ≈5% water

    Description

    Applications
    In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit