Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-D-tryptophan pentafluorophenyl ester, 97%

Click to view available options
Quantity:
250mg
1g
5g
136554-94-4
C32H21F5N2O4
592.522
MFCD00155634
KLPGTAYCHANVFY-XMMPIXPASA-N
fmoc-d-trp-opfp,d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester,d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester,2,3,4,5,6-pentafluorophenyl 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1h-indol-3-yl propanoate
53229920
(2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-indol-3-yl)propanoate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CNC5=CC=CC=C54)C(=O)OC6=C(C(=C(C(=C6F)F)F)F)F
136554-94-4
C32H21F5N2O4
592.522
MFCD00155634
KLPGTAYCHANVFY-XMMPIXPASA-N
fmoc-d-trp-opfp,d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester,d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester,2,3,4,5,6-pentafluorophenyl 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1h-indol-3-yl propanoate
53229920
(2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-indol-3-yl)propanoate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CNC5=CC=CC=C54)C(=O)OC6=C(C(=C(C(=C6F)F)F)F)F
Formula Weight 592.52
Physical Form Solid
Percent Purity 97%
Quantity 5g
Chemical Name or Material N-Fmoc-D-tryptophan pentafluorophenyl ester

storageNote1 : 2° to 8°C