Learn More
Aat BioquestSupplier Diversity Partner N-FMOC-ethylenediamine  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 4016
N-FMOC-ethylenediamine is a monoprotected diamine that is widely used as a building block in bioconjugate chemistry. It has been used in both oligonucleotide and peptide synthesis.
Catalog No. 50-195-3227