Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-glycyl-L-valine, 95%

Click to view available options
Quantity:
250mg
1g
5g
86895-14-9
C22H24N2O5
396.443
MFCD00190884
ORVHFHUXDLSLMX-FQEVSTJZSA-N
fmoc-gly-val-oh,l-valine, n-n-9h-fluoren-9-ylmethoxy carbonyl glycyl-9ci,fmoc-gly-val-oh,2s-2-2-9h-fluoren-9-ylmethoxy carbonyl amino acetamido-3-methylbutanoic acid
7019073
(2S)-2-[[2-(9H-fluoren-9-ylmethoxycarbonylamino)acetyl]amino]-3-methylbutanoic acid
CC(C)C(C(=O)O)NC(=O)CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
86895-14-9
C22H24N2O5
396.443
MFCD00190884
ORVHFHUXDLSLMX-FQEVSTJZSA-N
fmoc-gly-val-oh,l-valine, n-n-9h-fluoren-9-ylmethoxy carbonyl glycyl-9ci,fmoc-gly-val-oh,2s-2-2-9h-fluoren-9-ylmethoxy carbonyl amino acetamido-3-methylbutanoic acid
7019073
(2S)-2-[[2-(9H-fluoren-9-ylmethoxycarbonylamino)acetyl]amino]-3-methylbutanoic acid
CC(C)C(C(=O)O)NC(=O)CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Beilstein 6829030
Formula Weight 396.44
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material N-Fmoc-glycyl-L-valine