Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-alanine N-succinimidyl ester, 95%

Click to view available options
Quantity:
1g
5g
25g
73724-40-0
C22H20N2O6
408.41
MFCD00065613
FZBMNXOJLBTQHV-ZDUSSCGKSA-N
fmoc-ala-osu,fmoc-alanine-osu,fomc-ala-osu,fmoc-l-ala-osu,ambotzfaa6380,n-9h-fluorene-9-ylmethoxycarbonyl alanine succinimidyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-l-alanine succinimidyl ester,2,5-dioxopyrrolidin-1-yl n-9h-fluoren-9-yl methoxy carbonyl-l-alaninate,l-alanine,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,5-dioxo-1-pyrrolidinyl ester,2,5-dioxopyrrolidin-1-yl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoate
13670100
(2,5-dioxopyrrolidin-1-yl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoate
CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
73724-40-0
C22H20N2O6
408.41
MFCD00065613
FZBMNXOJLBTQHV-ZDUSSCGKSA-N
fmoc-ala-osu,fmoc-alanine-osu,fomc-ala-osu,fmoc-l-ala-osu,ambotzfaa6380,n-9h-fluorene-9-ylmethoxycarbonyl alanine succinimidyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-l-alanine succinimidyl ester,2,5-dioxopyrrolidin-1-yl n-9h-fluoren-9-yl methoxy carbonyl-l-alaninate,l-alanine,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,5-dioxo-1-pyrrolidinyl ester,2,5-dioxopyrrolidin-1-yl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoate
13670100
(2,5-dioxopyrrolidin-1-yl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoate
CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Beilstein 5462746
Formula Weight 408.41
Physical Form Powder
Percent Purity 95%
Color White
Quantity 25g
Chemical Name or Material N-Fmoc-L-alanine N-succinimidyl ester

storageNote1 : -30° to -10°C