Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-alaninol, 95%

Click to view available options
Quantity:
1g
5g
161529-13-1
C18H19NO3
297.354
MFCD00235913
GIZCEJUGNDJXMH-LBPRGKRZSA-N
fmoc-l-alaninol,fmoc-alaninol,s-9h-fluoren-9-yl methyl 1-hydroxypropan-2-yl carbamate,9h-fluoren-9-ylmethyl n-2s-1-hydroxypropan-2-yl carbamate,fmoc-dl-alaninol,fmoc-ala-ol,n-fmoc-l-alaninol,n-alpha-fmoc-l-alaninol,n-9-fluorenylmethoxycarbonyl-l-alaninol,9h-fluoren-9-yl methyl 1-hydroxypropan-2-yl carbamate
6924106
9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
CC(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
161529-13-1
C18H19NO3
297.354
MFCD00235913
GIZCEJUGNDJXMH-LBPRGKRZSA-N
fmoc-l-alaninol,fmoc-alaninol,s-9h-fluoren-9-yl methyl 1-hydroxypropan-2-yl carbamate,9h-fluoren-9-ylmethyl n-2s-1-hydroxypropan-2-yl carbamate,fmoc-dl-alaninol,fmoc-ala-ol,n-fmoc-l-alaninol,n-alpha-fmoc-l-alaninol,n-9-fluorenylmethoxycarbonyl-l-alaninol,9h-fluoren-9-yl methyl 1-hydroxypropan-2-yl carbamate
6924106
9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
CC(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Quantity 1g
Formula Weight 297.35
Physical Form Solid
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-alaninol

storageNote1 : -30° to -10°C