Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-aspartic acid 1-allyl ester, 97%

Click to view available options
Quantity:
250mg
1g
5g
144120-53-6
C22H21NO6
395.411
MFCD00467715
ZJMVIWUCCRKNHY-IBGZPJMESA-N
fmoc-asp-oall,fmoc-l-aspartic acid 1-allyl ester,s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid,3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid,fmocasp oh oallyl,ambotzfaa1355,fmoc-asp-allyl oh,fmoc-asp oh-oallyl,1-allyl n-fmoc-l-aspartate,fmoc-l-aspartic acid a-allyl ester
7020604
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid
C=CCOC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
144120-53-6
C22H21NO6
395.411
MFCD00467715
ZJMVIWUCCRKNHY-IBGZPJMESA-N
fmoc-asp-oall,fmoc-l-aspartic acid 1-allyl ester,s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid,3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid,fmocasp oh oallyl,ambotzfaa1355,fmoc-asp-allyl oh,fmoc-asp oh-oallyl,1-allyl n-fmoc-l-aspartate,fmoc-l-aspartic acid a-allyl ester
7020604
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid
C=CCOC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Beilstein 5460108
Formula Weight 395.41
Physical Form Powder
Percent Purity 97%
Color White
Quantity 250mg
Chemical Name or Material N-Fmoc-L-aspartic acid 1-allyl ester

storageNote1 : 2° to 8°C