Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-aspartic acid 1-benzyl ester, 95%

Click to view available options
Quantity:
250mg
1g
5g
86060-83-5
C26H23NO6
445.471
MFCD00198201
CBZSVHFNEMONDZ-QHCPKHFHSA-N
fmoc-asp-obzl,fmoc-l-aspartic acid alpha-benzyl ester,s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid,fmoc-l-aspartic acid-1-benzyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid alpha-benzyl ester,3s-4-benzyloxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid,fmoc-l-asp-obzl,fmoc-aspartic acid-obzl,pubchem14955,fmoc-l-aspartic,a-benzyl ester
11224591
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-phenylmethoxybutanoic acid
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
86060-83-5
C26H23NO6
445.471
MFCD00198201
CBZSVHFNEMONDZ-QHCPKHFHSA-N
fmoc-asp-obzl,fmoc-l-aspartic acid alpha-benzyl ester,s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid,fmoc-l-aspartic acid-1-benzyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid alpha-benzyl ester,3s-4-benzyloxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid,fmoc-l-asp-obzl,fmoc-aspartic acid-obzl,pubchem14955,fmoc-l-aspartic,a-benzyl ester
11224591
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-phenylmethoxybutanoic acid
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Beilstein 4606552
Formula Weight 445.47
Physical Form Powder
Percent Purity 95%
Color White
Quantity 250mg
Chemical Name or Material N-Fmoc-L-aspartic acid 1-benzyl ester

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C