Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-aspartic acid 4-cyclohexyl ester, 95%

Click to view available options
Quantity:
5g
25g
130304-80-2
C25H27NO6
437.492
MFCD00135420
OCMZRMNYEXIKQI-QFIPXVFZSA-N
fmoc-asp ochex-oh,fmoc-asp ochx-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid beta-cyclohexyl ester,fmoc-l-asp chx-oh,ambotzfaa1705,fmoc-l-asp ochx-oh,pubchem18985,fmoc-aspartic acid ochex,fmoc-l-aspartic acid beta-cyclohexyl ester
45072192
(2S)-4-cyclohexyloxy-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxobutanoic acid
C1CCC(CC1)OC(=O)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
130304-80-2
C25H27NO6
437.492
MFCD00135420
OCMZRMNYEXIKQI-QFIPXVFZSA-N
fmoc-asp ochex-oh,fmoc-asp ochx-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid beta-cyclohexyl ester,fmoc-l-asp chx-oh,ambotzfaa1705,fmoc-l-asp ochx-oh,pubchem18985,fmoc-aspartic acid ochex,fmoc-l-aspartic acid beta-cyclohexyl ester
45072192
(2S)-4-cyclohexyloxy-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxobutanoic acid
C1CCC(CC1)OC(=O)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 437.49
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material N-Fmoc-L-aspartic acid 4-cyclohexyl ester

storageNote1 : -30° to -10°C