Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-aspartic acid 4-methylpentyl ester, 98%

Click to view available options
Quantity:
250mg
1g
5g
180675-08-5
C25H29NO6
439.508
MFCD10001333
SYGKUYKLHYQKPL-NRFANRHFSA-N
fmoc-asp ompe-oh,l-aspartic acid,n-9h-fluoren-9-ylmethoxy carbonyl-, 4-1-ethyl-1-methylpropyl ester,n-9h-fluoren-9-ylmethoxycarbonyl-l-aspartic acid 4-1-methyl-1-ethylpropyl ester,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid non-preferred name,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid
56777270
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(3-methylpentan-3-yloxy)-4-oxobutanoic acid
CCC(C)(CC)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
180675-08-5
C25H29NO6
439.508
MFCD10001333
SYGKUYKLHYQKPL-NRFANRHFSA-N
fmoc-asp ompe-oh,l-aspartic acid,n-9h-fluoren-9-ylmethoxy carbonyl-, 4-1-ethyl-1-methylpropyl ester,n-9h-fluoren-9-ylmethoxycarbonyl-l-aspartic acid 4-1-methyl-1-ethylpropyl ester,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid non-preferred name,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid
56777270
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(3-methylpentan-3-yloxy)-4-oxobutanoic acid
CCC(C)(CC)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Formula Weight 439.51
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material N-Fmoc-L-aspartic acid 4-methylpentyl ester

storageNote1 : -30° to -10°C