Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, 97%

Click to view available options
Quantity:
1g
5g
86061-01-0
C29H24F5NO6
577.504
MFCD00037633
DWYWJUBBXKYAMY-IBGZPJMESA-N
fmoc-asp otbu-opfp,n-fmoc-beta-tert-butyl-l-aspartic acid pentafluorophenyl ester,fmoc-asp otbu-opfp,n-fmoc-l-aspartic acid 4-tert-butyl ester pentafluorophenyl ester,n-,a-fmoc-l-aspartic acid,a-tert-butyl ester pentafluorophenyl ester,n-alpha-fmoc-l-aspartic acid beta-tert-butyl ester pentafluorophenyl ester,n-9h-fluorene-9-ylmethoxycarbonyl-l-aspartic acid 1-pentafluorophenyl 4-tert-butyl ester,1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino butanedioate,n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid beta-t-butyl ester pentafluorphenyl ester
11548733
4-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioate
CC(C)(C)OC(=O)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
86061-01-0
C29H24F5NO6
577.504
MFCD00037633
DWYWJUBBXKYAMY-IBGZPJMESA-N
fmoc-asp otbu-opfp,n-fmoc-beta-tert-butyl-l-aspartic acid pentafluorophenyl ester,fmoc-asp otbu-opfp,n-fmoc-l-aspartic acid 4-tert-butyl ester pentafluorophenyl ester,n-,a-fmoc-l-aspartic acid,a-tert-butyl ester pentafluorophenyl ester,n-alpha-fmoc-l-aspartic acid beta-tert-butyl ester pentafluorophenyl ester,n-9h-fluorene-9-ylmethoxycarbonyl-l-aspartic acid 1-pentafluorophenyl 4-tert-butyl ester,1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino butanedioate,n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid beta-t-butyl ester pentafluorphenyl ester
11548733
4-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioate
CC(C)(C)OC(=O)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Beilstein 4222987
Formula Weight 577.5
Physical Form Powder
Percent Purity 97%
Color White
Quantity 1g
Chemical Name or Material N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester

storageNote1 : 2° to 8°C