Alfa Aesar™ N-Fmoc-L-beta-homophenylalanine, 95%

Manufacturer: Alfa Aesar™ H52052MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52052MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 193954-28-8
    Molecular Formula C25H23NO4
    Formula Weight 401.46
    MDL Number MFCD01863055
    Beilstein 7726557
    Synonym (S)-3-(Fmoc-amino)-4-phenylbutyric acid; Fmoc-beta-Homophe-OH
    Chemical Name or Material N-Fmoc-L-beta-homophenylalanine

    Specifications

    Formula Weight 401.46
    MDL Number MFCD01863055
    Quantity 250mg
    Molecular Formula C25H23NO4
    CAS 193954-28-8
    Beilstein 7726557
    Synonym (S)-3-(Fmoc-amino)-4-phenylbutyric acid; Fmoc-beta-Homophe-OH
    Chemical Name or Material N-Fmoc-L-beta-homophenylalanine
    Assay 95%

    Description

    Applications
    N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit