Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-glutamic acid 1-allyl ester, 98%

Click to view available options
Quantity:
250mg
1g
5g
144120-54-7
C23H23NO6
409.438
MFCD00467718
ORKKMGRINLTBPC-FQEVSTJZSA-N
fmoc-glu-oall,fmoc-l-glutamic acid 1-allyl ester,s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid,fmoc-l-glutamic acid,a-allyl ester,fmoc-glu-allyl ester,1-allyl n-fmoc-l-glutamate,n-fmoc-l-glutamic acid 1-allyl ester,fmoc-glu-oall hplc,1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate,n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester
7020606
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-prop-2-enoxypentanoic acid
C=CCOC(=O)C(CCC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
144120-54-7
C23H23NO6
409.438
MFCD00467718
ORKKMGRINLTBPC-FQEVSTJZSA-N
fmoc-glu-oall,fmoc-l-glutamic acid 1-allyl ester,s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid,fmoc-l-glutamic acid,a-allyl ester,fmoc-glu-allyl ester,1-allyl n-fmoc-l-glutamate,n-fmoc-l-glutamic acid 1-allyl ester,fmoc-glu-oall hplc,1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate,n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester
7020606
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-prop-2-enoxypentanoic acid
C=CCOC(=O)C(CCC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Beilstein 5460531
Formula Weight 409.44
Physical Form Powder
Percent Purity 98%
Color Brown
Melting Point 118°C to 122°C
Quantity 250mg
Chemical Name or Material N-Fmoc-L-glutamic acid 1-allyl ester

storageNote1 : 2° to 8°C